Merck
  • Apical Golgi localization of N,N'-diacetyllactosediamine synthase, beta4GalNAc-T3, is responsible for LacdiNAc expression on gastric mucosa.
Apical Golgi localization of N,N'-diacetyllactosediamine synthase, beta4GalNAc-T3, is responsible for LacdiNAc expression on gastric mucosa.
Glycobiology (2006-05-27)
Yuzuru Ikehara, Takashi Sato, Toru Niwa, Sachiko Nakamura, Masanori Gotoh, Sanae Kabata Ikehara, Katsue Kiyohara, Chihiro Aoki, Toshie Iwai, Hayao Nakanishi, Jun Hirabayashi, Masae Tatematsu, Hisashi Narimatsu
ABSTRACT
beta1,4-N-acetylgalactosaminyltransferase III (beta4GalNAc-T3), which was recently cloned and identified, exhibits GalNAc transferase activity toward a GlcNAcbeta residue with beta1,4-linkage, forming the N,N'-diacetyllactosediamine, GalNAcbeta1,4GlcNAc (LacdiNAc or LDN). Though LacdiNAc has not been found in the gastric mucosa, a large amount of transcript was detected in our previous study. To increase our knowledge of beta4GalNAc-T3 expression and its product LacdiNAc, we examined the exact localization of beta4GalNAc-T3 in human gastric mucosa using a newly developed antibody, monoclonal antibody (mAb) K1356. This antibody specifically detected the enzyme that transfected the beta4GalNAc-T3 gene into MKN45 cells, and the terminal betaGalNAc epitope yielded on the cell surface was recognized by a lectin, Wisteria floribunda agglutinin (WFA). beta4GalNAc-T3 was localized in the supra-nuclear region of surface mucous cells in gastric mucosa, and WFA positively stained the mucins secreted by the cells. In contrast, in the cells of the glandular compartment in the fundic glands and a few cells in the pyloric glands, beta4GalNAc-T3 was observed in the basolateral position of the nucleus, where no WFA reactivity was detected. The anti-Tn (GalNAcalpha-O-Ser/Thr) antibody staining did not overlap with the WFA staining. By measuring the binding activity of WFA using automated frontal affinity chromatography (FAC), we found WFA to bind most strongly LacdiNAc among the sugar chains examined. Neither beta4GalNAc-T3 nor WFA-positive staining was detected in intestinal metaplastic cells. These results suggest that the supra-nuclear expression of beta4GalNAc-T3 is essential for the formation of LacdiNAc on the surface mucous cells and that LacdiNAc and beta4GalNAc-T3 are novel differentiation markers of surface mucous cells in the gastric mucosa.