Merck
  • LC-MS/MS characterization of combined glycogenin-1 and glycogenin-2 enzymatic activities reveals their self-glucosylation preferences.
LC-MS/MS characterization of combined glycogenin-1 and glycogenin-2 enzymatic activities reveals their self-glucosylation preferences.
Biochimica et biophysica acta (2013-11-19)
Johanna Nilsson, Adnan Halim, Erik Larsson, Ali-Reza Moslemi, Anders Oldfors, Göran Larson, Jonas Nilsson
ABSTRACT
Glycogen synthesis is initiated by self-glucosylation of the glycosyltransferases glycogenin-1 and -2 that, in the presence of UDP-glucose, form both the first glucose-O-tyrosine linkage, and then stepwise add a series of α1,4-linked glucoses to a growing chain of variable length. Glycogen-1 and -2 coexist in liver glycogen preparations where the proteins are known to form homodimers, and they also have been shown to interact with each other. In order to study how glycogenin-1 and -2 interactions may influence each other's glucosylations we setup a cell-free expression system for in vitro production and glucosylation of glycogenin-1 and -2 in various combinations, and used a mass spectrometry based workflow for the characterization and quantitation of tryptic glycopeptides originating from glycogenin-1 and -2. The analysis revealed that the self-glucosylation endpoint was the incorporation of 4-8 glucose units on Tyr 195 of glycogenin-1, but only 0-4 glucose units on Tyr-228 of glycogenin-2. The glucosylation of glycogenin-2 was enhanced to 2-4 glucose units by the co-presence of enzymatically active glycogenin-1. Glycogenin-2 was, however, unable to glucosylate inactive glycogenin-1, at least not an enzymatically inactivated Thr83Met glycogenin-1 mutant, recently identified in a patient with severe glycogen depletion.