Merck
  • Diels-Alder chemistry of graphite and graphene: graphene as diene and dienophile.
Diels-Alder chemistry of graphite and graphene: graphene as diene and dienophile.
Journal of the American Chemical Society (2011-02-24)
Santanu Sarkar, Elena Bekyarova, Sandip Niyogi, Robert C Haddon
ABSTRACT
The zero-band-gap electronic structure of graphene enables it to function as either the diene or the dienophile in the Diels-Alder reaction, and this versatile synthetic method offers a powerful strategy for the reversible modification of the electronic properties of graphene under very mild conditions.
Sigma-Aldrich
2,3-Dimethoxy-1,3-butadiene, 95%