Merck
  • (E)-2,4-Dihydr-oxy-N'-(2-hydr-oxy-3-meth-oxy-5-nitro-benzyl-idene)benzohydrazide dihydrate.
(E)-2,4-Dihydr-oxy-N'-(2-hydr-oxy-3-meth-oxy-5-nitro-benzyl-idene)benzohydrazide dihydrate.
Acta crystallographica. Section E, Structure reports online (2010-01-01)
You-Yue Han, Yong-Hong Li, Qiu-Rong Zhao
ABSTRACT
The asymmetric unit of the title compound, C(15)H(13)N(3)O(7)·2H(2)O, consists of a hydrazone mol-ecule and two solvent water mol-ecules. The mol-ecule adopts an E configuration with respect to the C=N bond. It is relatively planar, with a dihedral angle between the two benzene rings of 2.6 (1)°. There are intra-molecular O-H⋯N and O-H⋯O hydrogen bonds in the hydrazone mol-ecule. In the crystal structure, symmetry-related mol-ecules are linked through inter-molecular N-H⋯O and O-H⋯O hydrogen bonds to form a three-dimensional network.
Sigma-Aldrich
3-Methoxy-5-nitrosalicylaldehyde, 98%