Merck
  • Enhancement of the photocatalytic activity of Ferrous Oxide by doping onto the nano-clinoptilolite particles towards photodegradation of tetracycline.
Enhancement of the photocatalytic activity of Ferrous Oxide by doping onto the nano-clinoptilolite particles towards photodegradation of tetracycline.
Chemosphere (2014-05-31)
Alireza Nezamzadeh-Ejhieh, Arezoo Shirzadi
ABSTRACT
Photodegradation of tetracycline (TC) aqueous solution by FeO doped onto nano-clinoptilolite particles was investigated using a high pressure Hg lamp as radiation source. Nano-particles of clinoptilolite were prepared using ball-milling of micro crystals of zeolite. The pretreated nano-particles ion exchanged in a ferrous solution and the Fe(II)-exchanged form was calcined at 450°C. All samples were characterized by FT-IR, DRS, SEM and XRD. The degradation extent was determined via UV-Vis absorption spectroscopy and COD. Based on the study of the effect of key operating parameters, the optimal conditions were determined to reach the higher efficiency of the process. The best photocatalytic activity was obtained in presence of the catalyst containing 5.4% FeO.
Sigma-Aldrich
Iron(II) oxide, −10 mesh, ≥99.6% trace metals basis
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
Tetracycline hydrochloride, ≥95% (European Pharmacopoeia HPLC assay)
Sigma-Aldrich
Tetracycline hydrochloride, powder
Sigma-Aldrich
Zeolite
Sigma-Aldrich
Tetracycline hydrochloride, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Aluminosilicate, mesostructured, MCM-41 (hexagonal)
Tetracycline hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Tetracycline hydrochloride, meets USP testing specifications
Supelco
Tetracycline hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Tetracycline hydrochloride, VETRANAL®, analytical standard