Merck
  • Separation of mandelic acid and its derivatives with new immobilized cellulose chiral stationary phase.
Separation of mandelic acid and its derivatives with new immobilized cellulose chiral stationary phase.
Journal of Zhejiang University. Science. B (2013-07-05)
Jie Zhou, Qian Liu, Guang-Jun Fu, Zhen-Zhong Zhang
ABSTRACT
A new liquid chromatographic method has been developed for the chiral separation of the enantiomers of mandelic acid and their derivatives 2-chloromandelic acid, 4-hydroxymandelic acid, 4-methoxymandelic acid, and 3,4,5-trismethoxymandelic acid. The enantiomers were separated by a CHIRALPAK(®) IC (250 mm×4.6 mm, 5 μm). Mandelic acid, 4-methoxymandelic acid, and 3,4,5-trismethoxymandelic acid were baseline resolved (resolution factor (RS)=2.21, RS=2.14, and RS=3.70, respectively). In contrast, the enantioselectivities between CHIRALPAK(®) IC and 2-chloromandelic acid and 4-hydroxymandelic acid investigated were low. By comparing the chromatographs of mandelic acid enantiomers and mandelic acid spiked with (R)-mandelic acid, it was determined that the first effluent was (R)-mandelic acid.