Merck
  • Ephedrine hydrochloride protects mice from LPS challenge by promoting IL-10 secretion and inhibiting proinflammatory cytokines.
Ephedrine hydrochloride protects mice from LPS challenge by promoting IL-10 secretion and inhibiting proinflammatory cytokines.
International immunopharmacology (2012-03-27)
Yuejuan Zheng, Ziyi Guo, Weigang He, Yang Yang, Yuhu Li, Aoxiang Zheng, Ping Li, Yan Zhang, Jinzhu Ma, Mingyue Wen, Muyi Yang, Huazhang An, Guang Ji, Yizhi Yu
ABSTRACT
Sepsis and its derivative endotoxic shock are still serious conditions with high mortality in the intensive care unit. The mechanisms that ensure the balance of proinflammatory cytokines and anti-inflammatory cytokine production are of particular importance. As an active α- and β-adrenergic agonist, ephedrine hydrochloride (EH) is a widely used agent for cardiovascular diseases, especially boosting blood pressure. Here we demonstrate that EH increased Toll-like receptor 4 (TLR4)-mediated production of interleukin 10 (IL-10) through p38 MAPK activation. Simultaneously, EH negatively regulated the production of proinflammatory cytokines. Consistently, EH increased lipopolysaccharide (LPS)-induced serum IL-10 and inhibited tumor necrotic factor-α (TNFα) production in vivo. As a result, EH treatment protected mice from endotoxic shock by lethal LPS challenge. In brief, our data demonstrated that EH could contribute to immune homeostasis by balancing the production of proinflammatory cytokines and anti-inflammatory cytokine in TLR4 signaling. This study provides a potential usage of EH in autoimmunologic diseases or other severe inflammations.
Sigma-Aldrich
DL-Ephedrine hydrochloride, 99%