Merck
  • Glucose metabolism activation by SHIP2 inhibitors via up-regulation of GLUT1 gene in L6 myotubes.
Glucose metabolism activation by SHIP2 inhibitors via up-regulation of GLUT1 gene in L6 myotubes.
European journal of pharmacology (2010-06-19)
Akira Suwa, Takeshi Kurama, Tadashi Yamamoto, Akihiko Sawada, Teruhiko Shimokawa, Ichiro Aramori
ABSTRACT
Lipid phosphatase SH2 domain-containing inositol 5'-phosphatase 2 (SHIP2) plays an important role in the regulation of insulin signaling. In this report, we identified AS1938909, a novel small-molecule SHIP2 inhibitor. AS1938909 showed potent inhibition of SHIP2 (Ki=0.44 microuM) and significant selectivity over other related phosphatases. Further, AS1938909 increased Akt phosphorylation, glucose consumption, and glucose uptake in L6 myotubes. Treatment of L6 myotubes with SHIP2 inhibitors for 48 h significantly induced expression of GLUT1 mRNA, but not that of GLUT4. These results suggest that pharmacological inhibition of SHIP2 activates glucose metabolism due, at least in part, to up-regulation of GLUT1 gene expression.
Sigma-Aldrich
AS1949490, ≥98% (HPLC)