Merck
π-Complexes from acyl cyanides and lithium dimethylcuprate(I).
Chemical communications (Cambridge, England) (2013-03-06)
Steven H Bertz, Richard A Hardin, Michael D Murphy, Craig A Ogle
ABSTRACT
Rapid injection of pyruvonitrile or benzoyl cyanide into solutions of Me2CuLi in THF-d8 at -100 °C gave complexes that were stable at this temperature. 1D NMR with multiply labelled substrates ((13)C/(15)N) and 2D NMR ((1)H/(13)C) identified them as the first cuprate-carbonyl π-complexes.
Sigma-Aldrich
Pyruvonitrile, 90%
Pricing and availability is not currently available.