Merck
Effects of daurisoline on intracellular Ca2+ activity in myocardium.
Zhongguo yao li xue bao = Acta pharmacologica Sinica (1996-05-01)
Z X Wang, J Q Zhu, F D Zeng, C J Hu, Y L Ma, S M Zhong
ABSTRACT
To explain the effect of daurisoline (DS) on delayed afterdepolarization (DAD). Ca(2+)-sensitive microelectrode technic was used to record intracellular Ca2+ activity (alpha Cai) and triggered activity (TA) arising from DAD in myocardium. Strophantin G 3 mumol.L-1 yielded an increase in resting myocardial alpha Cai by 0.19 +/- 0.11 mumol.L-1 and transient elevations of alpha Cai by 1.48 +/- 0.55 and 4.96 +/- 1.81 mumol.L-1, respectively during the development of DAD and TA. By pretreatment with DS or verapamil, strophantin G-caused elevations of the alpha Cai in resting and provoked myocardia were eliminated and TA disappeared. DS 50 mumol.L-1 reduced Na(+)-free medium-induced elevation of dog Purkinje fibrous alpha Cai and abolished caffeine-induced increase of dog myocardial alpha Cai. DS inhibited DAD and TA by preventing an increase of alpha Cai via transmembrane Ca2+ entry and Ca2+ release from the reticulum.
Sigma-Aldrich
Daurisoline, ≥98% (HPLC)