Merck
  • The effect of aging on acetaminophen pharmacokinetics, toxicity and Nrf2 in Fischer 344 rats.
The effect of aging on acetaminophen pharmacokinetics, toxicity and Nrf2 in Fischer 344 rats.
The journals of gerontology. Series A, Biological sciences and medical sciences (2013-07-19)
John Mach, Aniko Huizer-Pajkos, Victoria C Cogger, Catriona McKenzie, David G Le Couteur, Brett E Jones, Rafael de Cabo, Sarah N Hilmer
ABSTRACT
We investigated the effect of aging on hepatic pharmacokinetics and the degree of hepatotoxicity following a toxic dose of acetaminophen. Young and old male Fischer 344 rats were treated with 800 mg/kg acetaminophen (young n = 8, old n = 5) or saline (young n = 9, old n = 9). Serum measurements showed old rats treated with acetaminophen had significantly lower serum alanine aminotransferase and higher acetaminophen and acetaminophen glucuronide levels and creatinine, compared with acetaminophen treated young rats (p < .05). Immunoblotting and activity assays showed old saline-treated rats had twofold lower cytochrome P450 2E1 activity and threefold higher NAD(P)H quinone oxireductase 1 protein expression and activity than young saline-treated rats (p < .05), although Nrf2, glutathione cysteine ligase-modulatory subunit, glutathione cysteine ligase-catalytic subunit, and cytochrome P450 2E1 protein expressions were unchanged. Primary hepatocytes isolated from young rats treated with 10 mM acetaminophen had lower survival than those from old rats (52.4% ± 5.8%, young; 83.6% ± 1.7%, old, p < .05). The pharmacokinetic changes described may decrease susceptibility to acetaminophen-induced hepatotoxicity but may increase risk of nephrotoxicity in old age.
Product Description
Paracetamol, European Pharmacopoeia (EP) Reference Standard
Supelco
Paracetamol β-D-glucuronide, analytical standard
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, IgG fraction of antiserum, buffered aqueous solution
Supelco
Acetaminophen, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
USP
Acetaminophen, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetaminophen, analytical standard
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder
Supelco
Acetaminophen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®