Merck
  • Evaluation of amniotic membrane transplantation as an adjunct to medical therapy as compared with medical therapy alone in acute ocular burns.
Evaluation of amniotic membrane transplantation as an adjunct to medical therapy as compared with medical therapy alone in acute ocular burns.
Ophthalmology (2005-10-04)
Amol Tamhane, Rasik B Vajpayee, Nihar R Biswas, Ravindra M Pandey, Namrata Sharma, Jeewan S Titiyal, Radhika Tandon
ABSTRACT
To evaluate the role of amniotic membrane transplantation (AMT) in acute ocular burns. Prospective, randomized, controlled clinical trial. Patients with grade II to IV ocular burns within 3 weeks of injury were recruited. Thirty-seven patients, 7 of whom had bilateral involvement (total, 44 eyes), participated in the trial. Twenty eyes were included in group A (AMT) and 24 eyes were included in group B (controls). The eyes in the AMT group underwent AMT in addition to conventional medical therapy. In the control group, only conventional medical therapy was instituted. The patients' subjective assessments of relief of ocular discomfort, healing of the corneal epithelial defect, visual acuity, extent of corneal vascularization, formation of symblepharon, and tear function tests. At day 1, subjective ocular discomfort scores were reduced significantly in eyes with moderate burns in the AMT group compared with controls (P = 0.05), but there was no difference between the 2 groups in eyes with severe burns. The log mean percentage reduction in size of epithelial defect by day 7 was 7.43+/-0.89 after AMT and 6.23+/-1.10 with medical treatment alone in moderate grade burns at day 7 (P = 0.01), but there was no difference between the 2 groups in eyes with severe burns. There was no overall difference in the final visual acuity, symblepharon formation, corneal vascularization, and tear function tests between the 2 groups over the next 3 months and further follow-up. There was a high dropout rate for long-term follow-up. Amniotic membrane transplantation in eyes with acute ocular burns has advantages in terms of reduction of pain and promotion of early epithelialization in patients with moderate grade burns, but not so in severe burns. There seems to be no definite advantage to AMT over medical therapy alone in terms of improvement in visual acuity, appearance of symblepharon, corneal vascularization, and results of tear function tests on short-term follow-up.
Product Description
Homatropine hydrobromide, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.