Merck
  • Specific anti-tumor responses by cultured immune spleen cells. I. In vitro culture method and initial characterization of factors which block immune cell-mediated cytotoxicity in vitro.
Specific anti-tumor responses by cultured immune spleen cells. I. In vitro culture method and initial characterization of factors which block immune cell-mediated cytotoxicity in vitro.
International journal of cancer (1975-05-15)
K Nelson, S B Pollack, K E Hellström
ABSTRACT
Spleen cells from BALB/c mice which either bore syngeneic sarcomas or were normal controls were cultured in vitro. The culture supernatants of spleen cells from tumor-bearing mice inhibited (blocked) specific cell-mediated cytotoxicity to the tumor borne by the spleen donor. They also mediated specific antiserum-dependent cytotoxicity with control lymphoid cells. The appearance of blocking activity in culture supernatants was prevented by lysis of theta-positive spleen cells with antiserum and complement. The blocking activity was removed by passing culture supernatants through an anti-mouse immunoglobulin affinity column and was recovered in a 3 M NaSCN eluate of the column.
Sigma-Aldrich
Prealbumin from human plasma, lyophilized powder
Pricing and availability is not currently available.