Merck
  • The discovery and optimization of pyrimidinone-containing MCH R1 antagonists.
The discovery and optimization of pyrimidinone-containing MCH R1 antagonists.
Bioorganic & medicinal chemistry letters (2006-07-28)
Donald L Hertzog, Kamal A Al-Barazanji, Eric C Bigham, Michael J Bishop, Christy S Britt, David L Carlton, Joel P Cooper, Alex J Daniels, Dulce M Garrido, Aaron S Goetz, Mary K Grizzle, Yu C Guo, Anthony L Handlon, Diane M Ignar, Ronda O Morgan, Andrew J Peat, Francis X Tavares, Huiqiang Zhou
ABSTRACT
Optimization of a series of constrained melanin-concentrating hormone receptor 1 (MCH R1) antagonists has provided compounds with potent and selective MCH R1 activity. Details of the optimization process are provided and the use of one of the compounds in an animal model of diet-induced obesity is presented.
Sigma-Aldrich
Methyl 3-amino-2-thiophenecarboxylate, 99%
Pricing and availability is not currently available.