Merck
  • Organic base effects in NHC promoted O- to C-carboxyl transfer; chemoselectivity profiles, mechanistic studies and domino catalysis.
Organic base effects in NHC promoted O- to C-carboxyl transfer; chemoselectivity profiles, mechanistic studies and domino catalysis.
Organic & biomolecular chemistry (2011-04-21)
Craig D Campbell, Christopher J Collett, Jennifer E Thomson, Alexandra M Z Slawin, Andrew D Smith
ABSTRACT
The O- to C-carboxyl transfer of oxazolyl carbonates promoted by triazolinylidenes, generated in situ with NEt(3), shows a markedly different rate and chemoselectivity profile to the same reaction promoted by triazolinylidenes generated using KHMDS. The mechanism of these pathways has been probed through extensive crossover studies to understand this process. The use of NEt(3) as a base allows domino multi-step reaction sequences to be developed, although chiral NHCs only generate modest levels of asymmetric induction (<15% ee) in these domino reaction processes.