Merck
  • Preparation of (S)-1-Halo-2-octanols Using Ionic Liquids and Biocatalysts.
Preparation of (S)-1-Halo-2-octanols Using Ionic Liquids and Biocatalysts.
Molecules (Basel, Switzerland) (2009-11-20)
Mireia Oromí-Farrús, Jordi Eras, Núria Sala, Mercè Torres, Ramon Canela
ABSTRACT
Preparation of (S)-1-chloro-2-octanol and (S)-1-bromo-2-octanol was carried out by the enzymatic hydrolysis of halohydrin palmitates using biocatalysts. Halohydrin palmitates were prepared by various methods from palmitic acid and 1,2-octanediol. A tandem hydrolysis was carried out using lipases from Candida antarctica (Novozym 435), Rhizomucor miehei (Lipozyme IM), and "resting cells" from a Rhizopus oryzae strain that was not mycotoxigenic. The influence of the enzyme and the reaction medium on the selective hydrolysis of isomeric mixtures of halohydrin esters is described. Novozym 435 allowed preparation of (S)-1-chloro-2-octanol and (S)-1-bromo-2-octanol after 1-3 h of reaction at 40 degrees C in [BMIM][PF(6)].
Sigma-Aldrich
1,2-Octanediol, 98%
Pricing and availability is not currently available.