Merck
  • Lewis acid double metal cyanide catalysts for hydroamination of phenylacetylene.
Lewis acid double metal cyanide catalysts for hydroamination of phenylacetylene.
Chemical communications (Cambridge, England) (2011-03-08)
Annelies Peeters, Pieterjan Valvekens, Frederik Vermoortele, Rob Ameloot, Christine Kirschhock, Dirk De Vos
ABSTRACT
Double metal cyanides (DMCs) are highly active recyclable heterogeneous catalysts for hydroamination of phenylacetylene with 4-isopropylaniline. The best hydroamination yields are obtained with Zn-Co DMCs, especially if the particle size is decreased by a reverse emulsion synthesis technique.
Sigma-Aldrich
4-Isopropylaniline, 99%