Merck
  • Involvement of SMARCA2/BRM in the SWI/SNF chromatin-remodeling complex in schizophrenia.
Involvement of SMARCA2/BRM in the SWI/SNF chromatin-remodeling complex in schizophrenia.
Human molecular genetics (2009-04-14)
Minori Koga, Hiroki Ishiguro, Saori Yazaki, Yasue Horiuchi, Makoto Arai, Kazuhiro Niizato, Shuji Iritani, Masanari Itokawa, Toshiya Inada, Nakao Iwata, Norio Ozaki, Hiroshi Ujike, Hiroshi Kunugi, Tsukasa Sasaki, Makoto Takahashi, Yuichiro Watanabe, Toshiyuki Someya, Akiyoshi Kakita, Hitoshi Takahashi, Hiroyuki Nawa, Christian Muchardt, Moshe Yaniv, Tadao Arinami
ABSTRACT
Chromatin remodeling may play a role in the neurobiology of schizophrenia and the process, therefore, may be considered as a therapeutic target. The SMARCA2 gene encodes BRM in the SWI/SNF chromatin-remodeling complex, and associations of single nucleotide polymorphisms (SNPs) to schizophrenia were found in two linkage disequilibrium blocks in the SMARCA2 gene after screening of 11 883 SNPs (rs2296212; overall allelic P = 5.8 x 10(-5)) and subsequent screening of 22 genes involved in chromatin remodeling (rs3793490; overall allelic P = 2.0 x 10(-6)) in a Japanese population. A risk allele of a missense polymorphism (rs2296212) induced a lower nuclear localization efficiency of BRM, and risk alleles of intronic polymorphisms (rs3763627 and rs3793490) were associated with low SMARCA2 expression levels in the postmortem prefrontal cortex. A significant correlation in the fold changes of gene expression from schizophrenic prefrontal cortex (from the Stanley Medical Research Institute online genomics database) was seen with suppression of SMARCA2 in transfected human cells by specific siRNA, and of orthologous genes in the prefrontal cortex of Smarca2 knockout mice. Smarca2 knockout mice showed impaired social interaction and prepulse inhibition. Psychotogenic drugs lowered Smarca2 expression while antipsychotic drugs increased it in the mouse brain. These findings support the existence of a role for BRM in the pathophysiology of schizophrenia.