Merck
  • Cloning, expression and characterization of an extracellular enolase from Leuconostoc mesenteroides.
Cloning, expression and characterization of an extracellular enolase from Leuconostoc mesenteroides.
FEMS microbiology letters (2006-06-01)
Jin-Ha Lee, Hee-Kyoung Kang, Young-Hwan Moon, Dong Lyun Cho, Doman Kim, Jun-Yong Choe, R Honzatko, John F Robyt
ABSTRACT
Enolase on the surface of streptococci putatively facilitates pathogenic invasion of the host organisms. The related Leuconostoc mesenteroides 512FMCM is nonpathogenic, but it too has an extracellular enolase. Purified isolates of extracellular dextransucrase from cultures of L. mesenteroides contain minute amounts of enolase, which separate as small crystals. Expression of L. mesenteroides enolase in Escherichia coli provides a protein (calculated subunit mass of 47 546 Da) catalyzing the conversion of 2-phsopho-D-glycerate to phosphoenolpyruvate. The pH optimum is 6.8, with Km and kcat values of 2.61 mM and 27.5 s(-1), respectively. At phosphate concentrations of 1 mM and below, fluoride is a noncompetitive inhibitor with respect to 2-phospho-D-glycerate, but in the presence of 20 mM phosphate, fluoride becomes a competitive inhibitor. Recombinant enolase significantly inhibits the activity of purified dextransucrase, and does not bind human plasminogen. Results here suggest that in some organisms enolase may participate in protein interactions that have no direct relevance to pathogenic invasion.
Sigma-Aldrich
D(+)-2-Phosphoglyceric acid sodium salt hydrate, ≥75% (calc. on dry substance, enzymatic)
Pricing and availability is not currently available.