Merck
  • Structure of vulnibactin, a new polyamine-containing siderophore from Vibrio vulnificus.
Structure of vulnibactin, a new polyamine-containing siderophore from Vibrio vulnificus.
Biometals : an international journal on the role of metal ions in biology, biochemistry, and medicine (1994-04-01)
N Okujo, M Saito, S Yamamoto, T Yoshida, S Miyoshi, S Shinoda
ABSTRACT
A new siderophore named vulnibactin has been isolated from low iron cultures of Vibrio vulnificus, a human pathogen. The structure was established as N-[3-(2,3-dihydroxybenzamido)propyl]-1,3-bis[2-(2-hydroxy-phenyl)- trans-5 - methyl-2-oxazoline-4-carboxamido]propane by a combination of acid hydrolysis, nuclear magnetic resonance spectroscopy and positive fast atom bombardment mass spectrometry. Vulnibactin is characterized as containing one residue of 2,3-dihydroxybenzoic acid as well as two residues of salicylic acid, both of which are involved in the formation of oxazoline rings with L-threonine bound to a norspermidine backbone. In addition, two other compounds with siderophore activity were purified and their structures were also determined. These two compounds provided further support for the structure of vulnibactin.
Sigma-Aldrich
2,3-Dihydroxybenzoic acid, 99%