Merck
  • Oxidation of Thianthrene by the Ligninase of Phanerochaete chrysosporium.
Oxidation of Thianthrene by the Ligninase of Phanerochaete chrysosporium.
Applied and environmental microbiology (1988-07-01)
R P Schreiner, S E Stevens, M Tien
ABSTRACT
The oxidation of heterocyclic sulfur compounds reported to be part of the macrostructure of coal and petroleum was investigated. The oxidation of thianthrene solubilized in 10% dimethylformamide to thianthrene monosulfoxide in the presence of hydrogen peroxide was catalyzed by the ligninase from Phanerochaete chrysosporium.
Sigma-Aldrich
Thianthrene, 97%