Merck
  • Supplementation with mixed tocopherols increases serum and blood cell gamma-tocopherol but does not alter biomarkers of platelet activation in subjects with type 2 diabetes.
Supplementation with mixed tocopherols increases serum and blood cell gamma-tocopherol but does not alter biomarkers of platelet activation in subjects with type 2 diabetes.
The American journal of clinical nutrition (2006-01-10)
Michael W Clarke, Natalie C Ward, Jason H Y Wu, Jonathan M Hodgson, Ian B Puddey, Kevin D Croft
ABSTRACT
Some studies have shown potential benefit of vitamin E on platelet function, but several clinical trials failed to show improved cardiovascular outcome with alpha-tocopherol supplementation. Gamma-tocopherol, a major dietary form of vitamin E, may have protective properties different from those of alpha-tocopherol. We compared the effects of supplementation with alpha-tocopherol (500 mg) and a gamma-tocopherol-rich compound (500 mg, containing 60% gamma-tocopherol) on serum and cellular tocopherol concentrations, urinary tocopherol metabolite excretion, and in vivo platelet activation in subjects with type 2 diabetes. Fifty-eight subjects were randomly assigned to receive either 500 mg alpha-tocopherol/d, 500 mg mixed tocopherols/d, or matching placebo. Serum, erythrocyte, and platelet tocopherol and urinary metabolite concentrations were measured at baseline and after the 6-wk intervention. Soluble CD40 ligand, urinary 11-dehydro-thromboxane B2, serum thromboxane B2, soluble P-selectin, and von Willebrand factor were measured as biomarkers of in vivo platelet activation. Serum alpha-tocopherol increased with both tocopherol treatments. Serum and cellular gamma-tocopherol increased 4-fold (P < 0.001) in the mixed tocopherol group, whereas red blood cell gamma-tocopherol decreased significantly after alpha-tocopherol supplementation. Excretion of alpha-carboxyethyl-hydroxychroman increased significantly after supplementation with alpha-tocopherol and mixed tocopherols. Excretion of gamma-carboxyethyl-hydroxychroman increased significantly after supplementation with mixed tocopherols and after that with alpha-tocopherol, which may reflect the displacement of gamma-tocopherol by alpha-tocopherol due to incorporation of the latter into lipoproteins in the liver. Neither treatment had any significant effect on markers of platelet activation. Supplementation with alpha-tocopherol decreased red blood cell gamma-tocopherol, whereas mixed tocopherols increased both serum alpha-tocopherol and serum and cellular gamma-tocopherol. Changes in serum tocopherol closely reflect changes in cellular concentrations of tocopherols after supplementation.
Product Description
Supelco
δ-Tocopherol, analytical standard
Sigma-Aldrich
(+)-δ-Tocopherol, ≥90%