Merck
  • Studies on osteoporosis IV. Effect of benzo[b]thiophene-2- and dibenzothiophene-4-carboxylic acid on heparin accelerated osteoporosis.
Studies on osteoporosis IV. Effect of benzo[b]thiophene-2- and dibenzothiophene-4-carboxylic acid on heparin accelerated osteoporosis.
Journal of medicine (1980-01-01)
J C Robin, S D Sharma, K Francis, M Rosenstein, R Moore, J A Vida, C C Thomas, J L Ambrus
ABSTRACT
Total skeletal calcium was determined in female mice with the aid of whole body neutron activation analysis. Three months treatment with heaprin produced significant osteoporosis in C3-H/St(Ha) mice but not in C57/BL6(J) mice. This was more pronounced in the younger (5-8 months) than in the older (15-18 months) animals. Two 2-thiophene carboxylic acid derivatives showed significant osteoporosis preventive activity.
Sigma-Aldrich
Thianaphthene-2-carboxylic acid, 98%