Merck
  • Analysis of the cis-trans isomerization kinetics of L-alanyl-L-proline by the elution-band relaxation method.
Analysis of the cis-trans isomerization kinetics of L-alanyl-L-proline by the elution-band relaxation method.
Journal of chromatography (1987-05-22)
R Hanai, A Wada
ABSTRACT
The "elution-band relaxation method" has been applied to the analysis of the cis-trans kinetics isomerization of the proline of L-Ala-L-Pro using reversed-phase liquid chromatography. A procedure suitable for cases where neither isomer can be injected separately is described.
Sigma-Aldrich
Ala-Pro hydrate
Pricing and availability is not currently available.