Merck
  • Metabolism of uridine and determination of liver ribonucleic acid synthesis in developing and adult mice.
Metabolism of uridine and determination of liver ribonucleic acid synthesis in developing and adult mice.
The International journal of biochemistry (1985-01-01)
C Engelbrecht, T Yngner
ABSTRACT
The metabolism of [5-3H]uridine and the incorporation of the precursor into liver RNA was studied in developing (13-day-old) and adult (45-day-old) mice. Different time-courses of labelling and increased amounts of labelled catabolic products of uridine were found in liver and blood of developing mice compared with adult animals. This is suggested to be a consequence of enlarged metabolite pools resulting from a lower total amount of uracil-degrading enzymes in the developing mice. The labelling of the uracil nucleotides was decreased in the developing liver. However, in spite of a lower specific radioactivity of UTP, the RNA-specific radioactivity of developing liver was increased compared with adult liver. Also the labelling of liver RNA with [6-14C]orotic acid was found to be increased in developing mice, thus indicating a higher rate of RNA synthesis in these animals. A more pronounced difference in liver RNA labelling between the developing and the adult mice obtained with the use of [14C]orotic acid than with [3H]uridine may suggest that the de novo pathway, relative to the salvage pathways, is more important in developing than in adult liver.
Sigma-Aldrich
3-Ureidopropionic acid, ≥98.0% (T)
Pricing and availability is not currently available.