Merck
  • [Comparison between icariin and genistein in osteogenic activity of marrow stromal cells].
[Comparison between icariin and genistein in osteogenic activity of marrow stromal cells].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2013-09-10)
Jian Zhou, Ke-Ming Chen, Bao-Feng Ge, Xiao-Ni Ma, Xiao-Yu Guo, Kui Cheng, Yu-Hai Gao, Li-Juan Yan, Wen-Gui Shi
ABSTRACT
To compare the effect of icariin and genistein in the osteogenic differentiation of rat bone marrow stromal cells (rBMSC). Rat marrow stromal cells were seperated in vitro, and the optimal concentration of genisten and icriin were screened. Genistein and icariin with the concentration of 1 x 10(-5) mol x L(-1) were adopted to intereven rBMSCs cultured in vitro. Alkaline phosphatase (ALP) was determined at 3, 6, 9, 12,15 d after intervention; calcified nodule was detected with alizarin red staining at 12 d; OXS, Runx-2, bone morphogenetic protein (BMP-2) and Collagen-I mRNA expression were observed with Real-time RT-PCR at 12, 24, 48, 72, 96 h. Genistein and icariin with the concentration of 1 x 10(-5) mol x L(-1) could increase the activity of ALP and the content of Ca, regulate OXS, BMP-2, Runx-2 and Collagen-I mRNA expression. Icariin showed a stronger effect in improving the osteogenic differentiation of rat bone marrow stromal cells than genistein.
Sigma-Aldrich
Phosphatase, Alkaline bovine, recombinant, expressed in Pichia pastoris, ≥4000 units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, buffered aqueous glycerol solution, ≥4,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, buffered aqueous glycerol solution, 20-50 units/mg protein (in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, buffered aqueous solution, ≥2,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from porcine kidney, lyophilized powder, ≥100 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from calf intestinal mucosa, suitable for enzyme immunoassay, solution (clear, colorless), ~2500 U/mg protein (~10 mg/ml)
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, lyophilized powder, ≥10 DEA units/mg solid
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, lyophilized powder, 30-60 units/mg protein (in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, ammonium sulfate suspension, 30-90 units/mg protein (modified Warburg-Christian, in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, ≥5,500 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, BioUltra, ≥5,700 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline shrimp, ≥900 DEA units/mL, buffered aqueous glycerol solution, recombinant, expressed in proprietary host