Merck
  • Research of influence and mechanism of combining exercise with diet control on a model of lipid metabolism rat induced by high fat diet.
Research of influence and mechanism of combining exercise with diet control on a model of lipid metabolism rat induced by high fat diet.
Lipids in health and disease (2013-02-22)
Chen Shaodong, Zhou Haihong, Lin Manting, Li Guohui, Zhao Zhengxiao, Zhang Y M
ABSTRACT
To investigate the influence and mechanism of combining exercise with diet control on a model of lipid metabolism rat induced by high fat diet. Twenty-four male Wistar rats were randomly divided into 3 groups of 8: normal, model and intervention. The model group and intervention group were fed with high fat diet, while the normal group received basal feed. From day 1, the intervention group was randomly given interventions such as swimming exercise and dietary restriction. The interventions duration were 28 days. At the end of the experiment, the levels of rats' body weight and liver weight were detected, the serum levels of total cholesterol (TC), high density lipoprotein cholesterol (HDL-C), low density lipoprotein cholesterol (LDL-C) and hepatic triglyceride content (TG) were detected by using biochemical assay, serum level of gastrin (GAS), motilin (MTL) were assayed by the enzyme linked immunosorbent assay (ELISA). Compared with the level of body weight and liver weight in the normal rats, body weight and liver weight in the rat of the model group were significantly increase (P<0.05 or P<0.01). Plasma concentrations of TC, LDL-C and hepatic TG in the model group were significantly increased compared with those in the normal group (P<0.05 or P<0.01). The contents of GAS, MTL, HDL-C in the model rats'plasma were significantly reduced compared with those of the normal group (P<0.05 or P<0.01). Compared with those in the model group, rats' body weight, liver weight, serum TC, LDL-C, and TG content of liver in the intervention group decreased significantly (P<0.05 or P<0.01). Meanwhile, serum content of GAS, MTL, HDL-C were significantly improved in the intervention rats compared to the model group. The action of combining exercise with diet control for lipid metabolism disorder might be related to regulation of GAS, MTL and other gastrointestinal hormones.
Sigma-Aldrich
Motilin porcine, ≥97% (HPLC)