Merck
  • Rapid downregulation of innate immune cells, interleukin-12 and interleukin-23 in generalized pustular psoriasis with infliximab in combination with acitretin.
Rapid downregulation of innate immune cells, interleukin-12 and interleukin-23 in generalized pustular psoriasis with infliximab in combination with acitretin.
Dermatology (Basel, Switzerland) (2013-01-25)
M M Tang, Z Spanou, H Tang, F Schibler, N Pelivani, N Yawalkar
ABSTRACT
Generalized pustular psoriasis (GPP) is a severe inflammatory disease characterized by recurrent eruptions of sterile pustules on erythematous skin. Although tumor necrosis factor (TNF) antagonists may lead to a rapid resolution of GPP, the mechanism of action of these agents remains to be investigated. Here, we sought to evaluate markers of immune response in the skin of a patient who experienced a rapid amelioration of GPP after treatment with infliximab and acitretin. A skin biopsy was obtained before and 72 h after initiation of treatment. Immunohistochemical stainings were performed to characterize alterations of the infiltrates, the apoptosis marker caspase 3 and key cytokines like TNFα, interleukin (IL)-12, IL-23 and the chemokine CXCL8/IL-8. Parallel with clinical improvement, a striking decline of neutrophils, myeloid and plasmacytoid dendritic cells, M1 macrophages and partly of CD4+ T cells was observed. There was no evidence of increased apoptosis mediated through the caspase-3 pathway. A marked reduction particularly of IL-12 and IL-23 and, to a lesser degree, of TNFα and CXCL8/IL-8 was observed. A swift clinical improvement of GPP by infliximab and acitretin is associated with a marked reduction particularly of innate and partially of the acquired immune cells as well as IL-12 and IL-23.
Product Description
USP
Acitretin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acitretin, ≥98.0% (HPLC)
Acitretin, European Pharmacopoeia (EP) Reference Standard