Merck
  • Fabrication of hollow and porous structured GdVO4:Dy3+ nanospheres as anticancer drug carrier and MRI contrast agent.
Fabrication of hollow and porous structured GdVO4:Dy3+ nanospheres as anticancer drug carrier and MRI contrast agent.
Langmuir : the ACS journal of surfaces and colloids (2013-01-04)
Xiaojiao Kang, Dongmei Yang, Ping'an Ma, Yunlu Dai, Mengmeng Shang, Dongling Geng, Ziyong Cheng, Jun Lin
ABSTRACT
Hollow and porous structured GdVO(4):Dy(3+) spheres were fabricated via a facile self-sacrificing templated method. The large cavity allows them to be used as potential hosts for therapeutic drugs, and the porous feature of the shell allows guest molecules to easily pass through the void space and surrounding environment. The samples show strong yellow-green emission of Dy(3+) (485 nm, (4)F(9/2) → (6)H(15/2); 575 nm, (4)F(9/2) → (6)H(13/2)) under UV excitation. The emission intensity of GdVO(4):Dy(3+) was weakened after encapsulation of anticancer drug (doxorubicin hydrochloride, DOX) and gradually restored with the cumulative released time of DOX. These hollow spheres were nontoxic to HeLa cells, while DOX-loaded samples led to apparent cytotoxicity as a result of the sustained release of DOX. ICP measurement indicates that free toxic Gd ions can hardly dissolate from the matrix. The endocytosis process of DOX-loaded hollow spheres is observed using confocal laser scanning microscopy (CLSM). Furthermore, GdVO(4):Dy(3+) hollow spheres can be used for T(1)-weighted magnetic resonance (MR) imaging. These results implicate that the luminescent GdVO(4):Dy(3+) spheres with hollow and porous structure are promising platforms for drug storage/release and MR imaging.
Sigma-Aldrich
Dysprosium, ingot, 99.9% trace rare earth metals basis
Sigma-Aldrich
Dysprosium, chips, 99.9% trace rare earth metals basis