Merck
  • Infrared photodetectors based on CVD-grown graphene and PbS quantum dots with ultrahigh responsivity.
Infrared photodetectors based on CVD-grown graphene and PbS quantum dots with ultrahigh responsivity.
Advanced materials (Deerfield Beach, Fla.) (2012-09-01)
Zhenhua Sun, Zhike Liu, Jinhua Li, Guo-An Tai, Shu-Ping Lau, Feng Yan
ABSTRACT
Infrared photodetectors based on single-layer CVD-grown graphene and PbS quantum dots, which are fabricated by solution processing, show ultrahigh responsivities of up to 10(7) A/W under infrared light illumination. The devices fabricated on flexible plastic substrates have excellent bending stability. The photoresponse is attributed to the field-effect doping in graphene films induced by negative charges generated in the quantum dots.
Sigma-Aldrich
Lead(II) sulfide, 99.9% trace metals basis