Merck
  • Secondary metabolites from the roots of Neolitsea daibuensis and their anti-inflammatory activity.
Secondary metabolites from the roots of Neolitsea daibuensis and their anti-inflammatory activity.
Journal of natural products (2011-12-14)
Su-Ling Wong, Hsun-Shuo Chang, Guei-Jane Wang, Michael Y Chiang, Hung-Yi Huang, Chu-Huang Chen, Shiow-Chwen Tsai, Chu-Hung Lin, Ih-Sheng Chen
ABSTRACT
Bioassay-guided fractionation of the roots of Neolitsea daibuensis afforded three new β-carboline alkaloids, daibucarbolines A-C (1-3), three new sesquiterpenoids, daibulactones A and B (4 and 5) and daibuoxide (6), and 20 known compounds. The structures of 1-6 were determined by spectroscopic analysis and single-crystal X-ray diffraction. Daibucarboline A (1), isolinderalactone (7), 7-O-methylnaringenin (8), and prunetin (9) exhibited moderate iNOS inhibitory activity, with IC₅₀ values of 18.41, 0.30, 19.55, and 10.50 μM, respectively.
Sigma-Aldrich
Prunetin, ≥98.0% (TLC)