Merck
  • Enantioselective, palladium-catalyzed α-arylation of N-Boc pyrrolidine: in situ react IR spectroscopic monitoring, scope, and synthetic applications.
Enantioselective, palladium-catalyzed α-arylation of N-Boc pyrrolidine: in situ react IR spectroscopic monitoring, scope, and synthetic applications.
The Journal of organic chemistry (2011-07-01)
Graeme Barker, Julia L McGrath, Artis Klapars, Darren Stead, George Zhou, Kevin R Campos, Peter O'Brien
ABSTRACT
A comprehensive study of the enantioselective Pd-catalyzed α-arylation of N-Boc pyrrolidine has been carried out. The protocol involves deprotonation of N-Boc pyrrolidine using s-BuLi/(-)-sparteine in TBME or Et(2)O at -78 °C, transmetalation with ZnCl(2) and Negishi coupling using Pd(OAc)(2), t-Bu(3)P-HBF(4) and the aryl bromide. This paper reports several new features including in situ React IR spectroscopic monitoring of the process; use of (-)-sparteine and the (+)-sparteine surrogate to access products with opposite configuration; development of a catalytic asymmetric lithiation-Negishi coupling reaction; extension to a wide range of heteroaromatic bromides; total synthesis of (R)-crispine A, (S)-nicotine and (S)-SIB-1508Y via short synthetic routes; and examples of α-vinylation of N-Boc pyrrolidine using vinyl bromides exemplified by the total synthesis of naturally occurring (+)-maackiamine (thus establishing its configuration as (R)). In this way, the full scope and limitations of the methodology are delineated.
Sigma-Aldrich
N-Boc-pyrrolidine, 97%
Pricing and availability is not currently available.