Merck
  • Design, synthesis and structure-activity relationship of functionalized tetrahydro-β-carboline derivatives as novel PDE5 inhibitors.
Design, synthesis and structure-activity relationship of functionalized tetrahydro-β-carboline derivatives as novel PDE5 inhibitors.
Archiv der Pharmazie (2011-03-09)
Nermin S Ahmed, Bernard D Gary, Hethar N Tinsley, Gary A Piazza, Stefan Laufer, Ashraf H Abadi
ABSTRACT
Starting from tadalafil as a template, a series of functionalized tetrahydro-β-carboline derivatives have been prepared and identified as novel potent and selective PDE5 inhibitors. Replacing the 3,4-methylenedioxyphenyl at position 6 of tadalafil, together with elongation of the N2-methyl substituent and manipulation of the stereochemical aspects of the two chiral carbons led to the identification of compound XXI, a highly potent PDE5 inhibitor (IC(50)  = 3 nM). Compound XXI was also highly selective for PDE5 versus PDE3B, PDE4B, and PDE11A, with a selectivity index of 52 and 235 towards PDE5 rather than PDE11 with both cAMP and cGMP as substrate, respectively.
Sigma-Aldrich
1,2,3,4-Tetrahydro-9H-pyrido[3,4-b]indole, 98%