Merck
  • Vanadocene arsenate complexes: evidence of super-hyperfine coupling with (75)As.
Vanadocene arsenate complexes: evidence of super-hyperfine coupling with (75)As.
Magnetic resonance in chemistry : MRC (2010-06-11)
Jan Honzícek, Jaromír Vinklárek, Pavel Kratochvíl, Jan Hrbác
ABSTRACT
The reaction of vanadocene dichloride with sodium arsenate gives Cp(2)V(O(2)AsO(2)H). This compound was identified on the basis of the super-hyperfine coupling observed in the solution EPR spectrum. The effect of substitution was studied on the ring-substituted and ansa-bridged compounds.