Merck
  • Lack of genotoxicity of cross-linked acrylate polymers in four short-term genotoxicity assays.
Lack of genotoxicity of cross-linked acrylate polymers in four short-term genotoxicity assays.
Environmental and molecular mutagenesis (1991-01-01)
E D Thompson, J L Seymour, M J Aardema, R A LeBoeuf, B L Evans, D B Cody
ABSTRACT
Three cross-linked polyacrylate polymers containing either methylenebis-acrylamide (MBA), trimethylolpropane triacrylate (TMPTA), or triallylamine (TAA) cross-linkers were tested for genotoxicity with the Salmonella mammalian microsome assay, the L5178Y mouse lymphoma TK +/- assay, the unscheduled DNA synthesis assay in primary cultures of rat hepatocytes, and the in vivo bone marrow cytogenetic assay. The results indicate that none of the three polymers was genotoxic in these assays.
Sigma-Aldrich
Triallylamine, 99%