Merck
  • Further analysis of the antinociceptive action caused by p-methoxyl-diphenyl diselenide in mice.
Further analysis of the antinociceptive action caused by p-methoxyl-diphenyl diselenide in mice.
Pharmacology, biochemistry, and behavior (2008-10-22)
Cristiano R Jesse, Joao B T Rocha, Cristina W Nogueira, Lucielli Savegnago
ABSTRACT
The objective of this study was to extend our previous findings by investigating in greater detail the mechanisms that might be involved in the antinociceptive action of p-methoxyl-diphenyl diselenide, (MeOPhSe)(2), in mice. The pretreatment with nitric oxide precursor, l-arginine (600 mg/kg, intraperitoneal, i.p.), reversed antinociception caused by (MeOPhSe)(2) (10 mg/kg, p.o.) or N(G)-nitro-l-arginine (l-NOARG, 75 mg/kg, i.p.) in the glutamate test. Ondansetron (0.5 mg/kg, i.p., a 5-HT(3) receptor antagonist) and SCH23390 (0.05 mg/kg, i.p.., a D(1) receptor antagonist) blocked the antinociceptive effect caused by (MeOPhSe)(2). Conversely, pindolol (1 mg/kg, i.p., a 5-HT(1A)/(1B) receptor/beta adrenoceptor antagonist), WAY 100635 (0.7 mg/kg, i.p., a selective 5-HT(1A) receptor antagonist), ketanserin (0.3 mg/kg, i.p., a selective 5-HT(2A) receptor antagonist), prazosin (0.15 mg/kg, i.p., an alpha(1)-adrenoreceptor antagonist), yohimbine (1.0 mg/kg, i.p., an alpha(2)-adrenoreceptor antagonist), sulpiride (5 mg/kg, i.p., a D(2) receptor antagonist), naloxone (1 mg/kg, i.p., a non-selective opioid receptor antagonist) and caffeine (3 mg/kg, i.p., a non-selective adenosine receptor antagonist) did not change the antinociceptive effect of (MeOPhSe)(2). (MeOPhSe)(2) significantly inhibited nociception induced by intraplantar (i.pl.) injection of bradykinin (10 nmol/paw) and Des-Arg(9)-bradykinin (10 nmol/paw, a B(1) receptor agonist). (MeOPhSe)(2) significantly inhibited phorbol myristate acetate (PMA, 0.03 mug/paw, a protein kinase C (PKC) activator)-induced licking response. These results indicate that (MeOPhSe)(2) produced antinociception in mice through mechanisms that involve an interaction with nitrergic system, 5-HT(3) and D(1) receptors. The antinociceptive effect is related to (MeOPhSe)(2) ability to interact with kinin B(1) and B(2) receptors and PKC pathway mediated mechanisms.
Sigma-Aldrich
Bradykinin Fragment 2-9, ≥97% (HPLC)
Pricing and availability is not currently available.