Merck
Perioral dermatitis.
Acta dermatovenerologica Croatica : ADC (2008-06-11)
Suzana Ljubojević, Jasna Lipozencić, Petra Turcić
ABSTRACT
Perioral dermatitis is an inflammatory facial skin disorder that predominantly affects women. It is rarely diagnosed in children. The etiology of perioral dermatitis is unknown; however, uncritical use of topical corticosteroids often precedes skin lesions. There is a written diagnostic work-up, differential diagnosis and treatment.
Product Description
Supelco
Pramoxine hydrochloride, analytical standard, ≥98%