- Perspectives of CCK antagonists in pancreatic research. Part II. Experimental studies.
Perspectives of CCK antagonists in pancreatic research. Part II. Experimental studies.
International journal of pancreatology : official journal of the International Association of Pancreatology (1991-09-01)
T Takács, A Pap
PMID1757727
ABSTRACT
In this article, the effects of different classes of cholecystokinin (CCK) receptor antagonists in CCK-related physiological processes of the pancreas have been discussed. Both glutaramic acid derivatives and natural (benzodiazepine) analogs are potent, competitive antagonists of peripheral CCK receptors. These compounds thus provide a powerful tool for investigating the physiological and pharmacological actions of CCK in the gastrointestinal system, and have already clarified the role of CCK in pancreatic secretion and trophism or growth.