Merck
  • Lifetimes and reactivities of some 1,2-didehydroazepines commonly used in photoaffinity labeling experiments in aqueous solutions.
Lifetimes and reactivities of some 1,2-didehydroazepines commonly used in photoaffinity labeling experiments in aqueous solutions.
Biochemistry (2006-01-13)
Mary S Rizk, Xiaofeng Shi, Matthew S Platz
ABSTRACT
The reactive 1,2-didehydroazepine (cyclic ketenimine) intermediates produced upon photolysis of phenyl azide, 3-hydroxyphenyl azide, 3-methoxyphenyl azide, and 3-nitrophenyl azide in water and in HEPES buffer were studied by laser flash photolysis techniques with UV-vis detection of the transient intermediates. The lifetimes of the 1,2-didehydroazepines were obtained along with the absolute rate constants of their reactions with typical amino acids, nucleosides, and other simple reagents present in a biochemical milieu. The nitro substituent greatly accelerates the bimolecular reactions of the cyclic ketenimines, and the 3-methoxy group greatly decelerates the absolute reactivity of 1,2-didehydroazepines. The intermediate produced by photolysis of 3-hydroxyphenyl azide is much more reactive than the intermediate produced by photolysis of 3-methoxyphenyl azide. We propose that the hydroxyl-substituted 1,2-didehydoazepines rapidly (<10 micros) tautomerize in water to form azepinones and much more rapidly than the corresponding 3-methoxy-substituted cyclic ketenimines undergo hydrolysis. Azepinones react more rapidly with nucleophiles than do methoxy-substituted 1,2-didehydroazepines and are the active species present upon the photolysis of 3-hydroxyphenyl azide in aqueous solution.
Sigma-Aldrich
Azidobenzene solution, ~0.5 M in 2-methyltetrahydrofuran, ≥95.0% (HPLC)
Sigma-Aldrich
Azidobenzene solution, ~0.5 M in tert-butyl methyl ether