Merck
  • Transnitrosation of nitrosothiols: characterization of an elusive intermediate.
Transnitrosation of nitrosothiols: characterization of an elusive intermediate.
Journal of the American Chemical Society (2005-01-13)
Laura L Perissinotti, Adrián G Turjanski, Darío A Estrin, Fabio Doctorovich
ABSTRACT
We report an investigation of the reaction between (S)-nitroso-l-cysteine ethyl ester and l-cysteine ethyl ester as a model of physiologically relevant transnitrosation processes. Our theoretical and experimental evidence clearly supports the existence of a nitroxyl disulfide intermediate in solution.
Sigma-Aldrich
L-Cysteine ethyl ester hydrochloride, 98%