Merck
  • Biotransformation of alpha-isomethylionone to 1-(2,6,6-trimethyl-2-cyclohexen-1-yl)propan-2-one.
Biotransformation of alpha-isomethylionone to 1-(2,6,6-trimethyl-2-cyclohexen-1-yl)propan-2-one.
Bioscience, biotechnology, and biochemistry (2004-06-01)
Susumu Ishizaki, Masamichi Itoh, Tsuyoshi Komai, Tsutomu Honda, Takeshi Kitahara
ABSTRACT
The main biodegradation product of (+/-)-alpha-isomethylionone (2) with standard activated sludge was characterized as (+/-)-1-(2,6,6-trimethyl-2-cyclohexen-1-yl)propan-2-one (1) by its analysis and synthesis. Both enantiomers (1a and 1b) of 1 were synthesized by starting from (R)- and (S)-2,4,4-trimethyl-2-cyclohexen-1-ol (3a and 3b), respectively.
Sigma-Aldrich
α-Cetone, odorant used in allergy studies
Pricing and availability is not currently available.