Merck
  • Importance of NF-kappaB in rheumatoid synovial tissues: in situ NF-kappaB expression and in vitro study using cultured synovial cells.
Importance of NF-kappaB in rheumatoid synovial tissues: in situ NF-kappaB expression and in vitro study using cultured synovial cells.
Annals of the rheumatic diseases (2001-06-19)
S Yamasaki, A Kawakami, T Nakashima, H Nakamura, M Kamachi, S Honda, Y Hirai, A Hida, H Ida, K Migita, Y Kawabe, T Koji, I Furuichi, T Aoyagi, K Eguchi
ABSTRACT
To examine whether inhibition of NF-kappaB induces apoptosis of human synovial cells stimulated by tumour necrosis factor alpha (TNFalpha), interleukin 1beta (IL1beta), and anti-Fas monoclonal antibody (mAb). The expression of proliferating cell nuclear antigen (PCNA), NF-kappaB, and the presence of apoptotic synovial cells were determined in synovial tissues. Apoptosis of cultured synovial cells was induced by inhibition of NF-kappaB nuclear translocation by Z-Leu-Leu-Leu-aldehyde (LLL-CHO). The activation of caspase-3 and expression of XIAP and cIAP2 in synovial cells in LLL-CHO induced apoptosis was also examined. Abundant PCNA+ synovial cells were found in rheumatoid arthritis (RA) synovial tissue, though a few apoptotic synovial cells were also detected in the RA synovial tissues. Nuclear NF-kappaB was expressed in RA synovial cells. Electrophoretic mobility shift assay showed that treatment of cells with TNFalpha or IL1beta significantly stimulated nuclear NF-kappaB activity. A small number of apoptotic synovial cells expressing intracellular active caspase-3 were found after treatment of cells with LLL-CHO. Although treatment of RA synovial cells with TNFalpha or IL1beta alone did not induce apoptosis, apoptosis induced by LLL-CHO and caspase-3 activation were clearly enhanced in TNFalpha or IL1beta stimulated synovial cells compared with unstimulated synovial cells. Furthermore, induction of apoptosis of synovial cells with caspase-3 activation by anti-Fas mAb was clearly increased by LLL-CHO. The expression of cIAP2 and XIAP in synovial cells may not directly influence the sensitivity of synovial cells to apoptosis induced by LLL-CHO. The results suggest that NF-kappaB inhibition may be a potentially important therapeutic approach for RA by correcting the imbalance between apoptosis and proliferation of synovial cells in RA synovial tissue.
Sigma-Aldrich
Leu-Leu-Leu, ≥90% (elemental analysis)