Merck
Determination of six pterins in urine by LC-MS/MS.
Bioanalysis (2012-08-11)
Gabriella Allegri, Heleno José Bezerra Costa Netto, Luiz Nelson Lopes Ferreira Gomes, Maria Lúcia Costa de Oliveira, Fernanda Bertão Scalco, Francisco Radler de Aquino Neto
ABSTRACT
The present work describes an analytical method for urinary pterins by LC-MS/MS, with emphasis on the separation of 6- and 7-positional isomers of bio- and neopterins. Urine sample preparation consisted of oxidation by MnO(2), filtration and direct dilution in the mobile phase. The method was validated in urine spiked at five concentration levels with true triplicates of each level. Separation of the pterins, including the positional isomers, was achieved by employing a LUNA amino column. Six pterins were quantified (pterin, isoxanthopterin, 6-biopterin, 7-biopterin, 6-neopterin, 7-neopterin) and a linear behavior was observed; LOD varied from 7 to 360 pg/ml and correlation coefficients above 0.98 were obtained for all pterins. In addition, pterin levels were evaluated in 41 urine samples of healthy subjects, in ten urine samples of patients with classical phenylketonuria (PKU) and in one with atypical PKU. The proposed method allowed to identify, separate and quantify six pterins in urine, using a simple and rapid sample preparation. The atypical PKU was unequivocally differentiated from the classical form, demonstrating that this method could be very useful for characterization and follow-up of diseases.