Merck
  • Influence of hydropower dams on the composition of the suspended and riverbank sediments in the Danube.
Influence of hydropower dams on the composition of the suspended and riverbank sediments in the Danube.
Environmental pollution (Barking, Essex : 1987) (2007-04-03)
Gerard Klaver, Bertil van Os, Philippe Negrel, Emmanuelle Petelet-Giraud
ABSTRACT
Large hydropower dams have major impacts on flow regime, sediment transport and the characteristics of water and sediment in downstream rivers. The Gabcikovo and Iron Gate dams divide the studied Danube transect (rkm 1895-795) into three parts. In the Gabcikovo Reservoir (length of 40km) only a part of the incoming suspended sediments were deposited. Contrary to this, in the much larger Iron Gate backwater zone and reservoir (length of 310km) all riverine suspended sediments were deposited within the reservoir. Subsequently, suspended sediments were transported by tributaries into the Iron Gate backwater zone. Here they were modified by fractional sedimentation before they transgressed downstream via the dams. Compared with undammed Danube sections, Iron Gate reservoir sediment and suspended matter showed higher clay contents and different K/Ga and Metal/Ga ratios. These findings emphasize the importance of reservoir-river sediment-fractionation.
Sigma-Aldrich
Sodium oxide, 80%
Pricing and availability is not currently available.