Merck
  • Phosphorylation of ethanolamine, methylethanolamine, and dimethylethanolamine by overexpressed ethanolamine kinase in NIH 3T3 cells decreases the co-mitogenic effects of ethanolamines and promotes cell survival.
Phosphorylation of ethanolamine, methylethanolamine, and dimethylethanolamine by overexpressed ethanolamine kinase in NIH 3T3 cells decreases the co-mitogenic effects of ethanolamines and promotes cell survival.
European journal of biochemistry (1998-05-13)
B Malewicz, J J Mukherjee, K S Crilly, W J Baumann, Z Kiss
ABSTRACT
Ethanolamine (Etn), as well as its N-methyl (MeEtn) and N,N-dimethyl (Me2Etn) analogues, were recently shown to potentiate the stimulatory effect of insulin on DNA synthesis in serum-starved NIH 3T3 fibroblasts. In the present work we assessed the impact of the co-mitogenic effects of Etn and its methyl analogues on cell proliferation and cell survival, and examined whether the cell growth regulatory effects of these ethanolamines involve an Etn-kinase-mediated phosphorylation step. For this purpose, NIH 3T3 sublines highly overexpressing Drosophila Etn kinase and an appropriate vector control line were utilized and the effects of Etn, MeEtn, Me2Etn, methylamine (MeNH2), and dimethylamine (Me2NH) were studied. 31P-NMR analysis of the water-soluble cell metabolites revealed that both MeEtn and Me2Etn, but not choline, are excellent substrates for the expressed Etn kinase. The methylated ethanolamines (MeEtn and Me2Etn) and methylamines (MeNH2, Me2NH) were used as Etn models that can or cannot be phosphorylated, respectively. In serum-starved vector control cells, both MeNH2 (1 mM) and Me2NH (1 mM) were more effective than Etn in enhancing insulin-induced DNA synthesis, and both were almost as effective as MeEtn and Me2Etn. However, in the Etn kinase overexpressor cells the potentiating effects of Etn, MeEtn and Me2Etn, but not those of MeNH2 and Me2NH, were significantly reduced. Moreover, in the overexpressor cells, lower concentrations of Etn (50-200 microM) inhibited the combined mitogenic effects of Me2NH (1 mM) and insulin. These data are consistent with a mechanism in which the phosphorylated and non-phosphorylated ethanolamines are negative and positive regulators of insulin-induced mitogenesis, respectively. After incubating the cells for 13 days in serum-free medium in 96-well microplates, there was a steady decrease in cell numbers in both cell lines. However, between 6-13 days, 0.1-1 mM MeEtn and, particularly, Me2Etn provided significant protection against cell death in the Etn kinase overexpressor cells. In vector control cells, only Me2Etn in combination with insulin had similar effects on cell survival. The data suggest that phosphorylated ethanolamines may function as promoters of cell survival.
Sigma-Aldrich
2-(Methylamino)ethanol, ≥98%