Merck
Acute ammonium dichromate poisoning.
Annals of emergency medicine (1994-10-01)
K L Meert, J Ellis, R Aronow, E Perrin
ABSTRACT
We report the ingestion of ammonium dichromate by a child that resulted in multiple-organ-system failure and death. Exchange transfusion and hemodialysis were ineffective in removing significant amounts of chromium or causing sustained clinical improvement. We suggest that immediate, large doses of the reducing agent ascorbic acid would allow effective reduction of hexavalent chromium with less cellular toxicity.
Sigma-Aldrich
Ammonium dichromate, ACS reagent, ≥99.5%