Merck
  • [Effect of various methods of resection of the stomach in liberation of cholecystokinin, neurotensin and on pancreatic function].
[Effect of various methods of resection of the stomach in liberation of cholecystokinin, neurotensin and on pancreatic function].
Zentralblatt fur Chirurgie (1995-01-01)
M Fuchs, H Köhler, A Schafmayer
ABSTRACT
In a follow up study 19 patients after Billroth-I, 22 patients after Billroth-II-resection and 38 patients after total gastrectomy (23 with Roux-Y-reconstruction and 15 with Longmire-Gütgemann) underwent a stool fat determination and an indirect pancreatic function test with fluorescein dilaurate in serum und urine. In 9 of 19 cases (47.3%) after B-I-resection and 14 of 22 patients (63.5%) after B-II-resection there were pathological results of the PLT-test in urine. After total gastrectomy as well with reconstruction of the duodenal passage as with its exclusion the PLT-test results were pathological in 60% resp. 87%. These results indicate a secondary pancreatic insufficiency following gastric resection.
Sigma-Aldrich
Fluorescein dilaurate, suitable for fluorescence, ≥97.0% (HPCE)
Pricing and availability is not currently available.