Merck
  • Mechanistic implications of the inhibition of peptidases by amino aldehydes and bestatin.
Mechanistic implications of the inhibition of peptidases by amino aldehydes and bestatin.
Biochimica et biophysica acta (1985-07-01)
L Frick, R Wolfenden
ABSTRACT
alpha-Amino aldehydes and bestatin are found to be effective inhibitors of a cytosolic dipeptidase (rat testicular peptidase C), and a cytosolic tripeptidase (rat kidney peptidase B, EC 3.4.11.4), as well as cytosolic leucine aminopeptidase (pig kidney peptidase S, EC 3.4.11.1). Aldehyde hydrates and bestatin share a resemblance to intermediates that might be formed during direct attack by water on peptide substrates, affording a possible explanation for their tight binding. Alternatively, inhibitors of both kinds might form derivatives of an active site nucleophile, resembling intermediates in a double-displacement mechanism. Exchange experiments with H218O suggest that bestatin is bound intact by leucine aminopeptidase, lending support to the first of these two mechanisms.
Sigma-Aldrich
1-Formylpyrrolidine, 97%