Merck
  • Ancymidol blocks trichothecene biosynthesis and leads to accumulation of trichodiene in Fusarium sporotrichioides and Gibberella pulicaris.
Ancymidol blocks trichothecene biosynthesis and leads to accumulation of trichodiene in Fusarium sporotrichioides and Gibberella pulicaris.
Applied and environmental microbiology (1987-08-01)
A E Desjardins, R D Plattner, M N Beremand
ABSTRACT
Ancymidol, a plant growth regulator, inhibited biosynthesis of diacetoxyscirpenol by Gibberella pulicaris (Fusarium sambucinum) in a defined liquid medium. Ancymidol also inhibited biosynthesis of T-2 toxin by a wild-type strain of Fusarium sporotrichioides and biosynthesis of diacetoxyscirpenol, deacetylated calonectrin, and dideacetylated calonectrin by mutant strains of this species. Ancymidol-treated cultures accumulated the hydrocarbon trichodiene, a biosynthetic precursor of the trichothecenes. Ancymidol did not block trichodiene accumulation by a trichodiene-producing mutant strain of F. sporotrichioides. Ancymidol appears to block the trichothecene biosynthetic pathway after formation of trichodiene and before formation of trichothecenes containing four or more oxygen atoms.
Sigma-Aldrich
Ancymidol, suitable for plant cell culture, BioReagent
Pricing and availability is not currently available.