- [Comparison of the effects of glutamate antibodies on neuronal activity of caspase 3 and memory impairment in rats induced by injection of Abeta(25-35) into the Meynert nuclei].
[Comparison of the effects of glutamate antibodies on neuronal activity of caspase 3 and memory impairment in rats induced by injection of Abeta(25-35) into the Meynert nuclei].
Patologicheskaia fiziologiia i eksperimental'naia terapiia (2013-06-29)
V V Kolobov, V G Fomina, V Yu Gorbatov, T V Davydova
PMID23805710
ABSTRACT
In experiments on rats showed that intranasal administration of glutamate antibodies in a dose of 300 microg/kg after 1 h after bilateral injection of neurotoxic fragment of beta-amyloid protein (25-35)--Abeta(25-35)--into the Meynert nuclei restores learning ability in the test of passive avoidance on 3 and 14 days of the experiment. Antibodies to glutamate decrease significantly increasing caspase 3 activity, detected on Day 3 after injection of Abeta(25-35), in samples of the prefrontal cortex and hippocampus but not hypothalamus. Intranasal administration of gamma-globulin had no effect on the performance of violations of mnestic functions and caspase 3 activity.
Product Description
Supelco
L-Glutamic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamic acid, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-100.5%